Would you like to log in again?
LB Agar Plate with Chloramphenicol, Thermo Scientific
Supplier: Thermo Scientific
J62923-EQF J61140-EQF J61140-EQF J62923-EQF
AAJ61140-EQFEA 158.88 USD InStock
AAJ61140-EQF CAAAJ62923-EQF CAAAJ61140-EQF AAJ62923-EQF
LB Agar Plate with Chloramphenicol, Thermo Scientific
Media for Microbiology Prepared Media for Microbiology
This Miller’s LB is a highly-referenced microbial growth medium used for the cultivation of <italic>E.coli</italic>.
It is used with recombinant strains of <italic>E.coli</italic> containing plasmids with the resistance gene to 25 µg/mL chloramphenicol such as pBluescript, pGEM, or the pUC series of plasmids.
Caution: Keep away from strong oxidizing agents and in a container tightly closed in a dry and well-ventilated place.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty