Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Leupeptin hemisulphate

Supplier: Thermo Scientific
J61188-MB J61188-MB J61188-MC J61188-MC
AAJ61188-MBEA 198.76 USD InStock
CAAAJ61188-MB AAJ61188-MB CAAAJ61188-MC AAJ61188-MC
Leupeptin hemisulphate
Proteins and Peptides
Powder

MDL No.: MFCD00037012

CAS No.: 103476-89-7
Molecular Formula: C20H38N6O4.0.5H2SO4
Formula Weight: 475.59
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water at 50mg/ml
MDL No.: MFCD00037012

Reversible protease inhibitor which inhibits cathepsin B, calpain and trypsin

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty