Synonyms:
Nucleic Acid
J61215-MC
J61215-ME
J61215-ME
J61215-03
J61215-MC
J61215-03
AAJ61215-03EA
693.85
USD
InStock
AAJ61215-03
CAAAJ61215-03
CAAAJ61215-ME
CAAAJ61215-MC
AAJ61215-ME
AAJ61215-MC
Ribonucleic acid (from bakers yeast)
Ribonucleic acid
Ribonucleic acid (RNA) from baker's yeast S. cerevisiae may be used as a substrate for studying ribonuclease activities of enzymes such as RNAase(s). It is also suitable for use as a carrier in nucleic acid purification and precipitation.
- Off-white to pale brown powder
- Soluble in water
- Store at cold temperature
Suitable for use as a ribonuclease substrate.
Caution: Not suitable for use as a carrier in nucleic acid purification and precipitation.