Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adrenocorticotropic Hormone (1-13)

Supplier: Thermo Scientific
J61247-LB0 J61247-MCR J61247-EXD J61247-LB0 J61247-EXD J61247-MCR
AAJ61247-EXDEA 80.86 USD InStock
CAAAJ61247-EXD AAJ61247-EXD CAAAJ61247-LB0 AAJ61247-LB0 CAAAJ61247-MCR AAJ61247-MCR
Adrenocorticotropic Hormone (1-13)
Proteins and Peptides
CAS No.: 22006-64-0
Molecular Formula: C75H106N20O19S
Formula Weight: 1623.85
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00133030

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty