Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vasopressin ≥98%

Supplier: Thermo Scientific Chemicals

J61248-MB J61248-LB0 J61248-MC J61248-MC J61248-MB
AAJ61248-MBEA 487.15 USD InStock
CAAAJ61248-LB0 AAJ61248-MB CAAAJ61248-MC AAJ61248-MC CAAAJ61248-MB
Vasopressin ≥98%
Vasopressin
Has antidiuretic activity and plays a role in water balance in animals

Lyophilized powder


Formula: C₄₆H₆₅N₁₅O₁₂S₂
MW: 1084.24 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00076738
CAS Number: 11000-17-2
EINECS: 234-236-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty