Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Paromomycin sulfate

Supplier: Thermo Scientific Chemicals
Danger

J61274-14 J61274-03 J61274-03 J61274-06 J61274-06 J61274-14
AAJ61274-03EA 55.14 USD InStock
AAJ61274-03 AAJ61274-14 AAJ61274-06 CAAAJ61274-06 CAAAJ61274-14 CAAAJ61274-03
Paromomycin sulfate
Paromomycin sulfate
Inhibits initiation and elongation during protein synthesis

White to off-white powder


Formula: C₂₃H₄₅N₅O₁₄·H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00079278
CAS Number: 1263-89-4
EINECS: 215-031-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty