Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+)-1-Deoxynojirimycin hydrochloride

Supplier: Thermo Scientific Chemicals
Synonyms: 1-Deoxymannojirimycin hydrochloride

J61277-LB0 J61277-MA J61277-MB J61277-MA J61277-LB0 J61277-MB
AAJ61277-LB0EA 171.38 USD InStock
AAJ61277-LB0 AAJ61277-MB CAAAJ61277-LB0 AAJ61277-MA CAAAJ61277-MA CAAAJ61277-MB
(+)-1-Deoxynojirimycin hydrochloride
(+)-1-Deoxynojirimycin hydrochloride
Selective inhibitor of a-mannosidase


Formula: C₆H₁₄ClNO₄
MW: 199.64 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00083611
CAS Number: 73465-43-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty