J61299-18
J61299-22
J61299-18
J61299-22
AAJ61299-22EA
114.97
USD
InStock
AAJ61299-22
CAAAJ61299-22
AAJ61299-18
CAAAJ61299-18
Streptomycin sulfate cell culture reagent
Streptomycin sulphate
Inhibits initiation and causes misreading of rRNA in protein synthesis.
Powder