Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Azauridine

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,2,4-triazine-3,5(2H,4H)-dione

J61309-03 J61309-06 J61309-03 J61309-06
AAJ61309-06EA 547.86 USD InStock
CAAAJ61309-03 CAAAJ61309-06 AAJ61309-06 AAJ61309-03
6-Azauridine
6-Azauridine
Powder


Formula: C₈H₁₁N₃O₆
MW: 245.19 g/mol
Melting Pt: 160…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006472
CAS Number: 54-25-1
EINECS: 200-199-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty