Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Butanediol dimethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Busulfan

J61348-06 J61348-06
AAJ61348-06EA 26.77 USD InStock
CAAAJ61348-06 AAJ61348-06
1,4-Butanediol dimethanesulfonate 98%
Busulfan
A cell cycle non-specific alkylating antineoplastic agent

Crystalline powder


Formula: C₆H₁₄O₆S₂
MW: 246.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007562
CAS Number: 55-98-1
EINECS: 200-250-2
UN: 2811
ADR: "6.1",III
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty