Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N''-Hydroxyguanidine acetate

Supplier: Thermo Scientific Chemicals

J61354-MF J61354-MF
AAJ61354-MFEA 305.33 USD InStock
AAJ61354-MF CAAAJ61354-MF
N''-Hydroxyguanidine acetate
N''-Hydroxyguanidine acetate
Reacts with nitric oxide forming an adduct which is a potent and stable vasodilator

Crystals


Formula: C₃H₉N₃O₃
Melting Pt: 132…134 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD22123289
CAS Number: 6345-29-5
EINECS: 228-749-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty