Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Iodo-2'-deoxycytidine 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidin-2(1H)-one

J61356-03 J61356-03 J61356-04 J61356-04
AAJ61356-04EA 399.73 USD InStock
CAAAJ61356-03 CAAAJ61356-04 AAJ61356-04 AAJ61356-03
5-Iodo-2'-deoxycytidine 99%
5-Iodo-2'-deoxycytidine
Powder


Formula: C₉H₁₂IN₃O₄
MW: 353.12 g/mol
Melting Pt: 180…183 °C
Storage Temperature: Freezer
MDL Number: MFCD00038063
CAS Number: 611-53-0
EINECS: 210-269-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty