Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Chlorokynurenic acid ≥99%

Supplier: Thermo Scientific Chemicals
Synonyms: 7-Chloro-4-hydroxyquinoline-2-carboxylic acid

J61359-MA J61359-MA J61359-MF J61359-MF
AAJ61359-MAEA 202.91 USD InStock
AAJ61359-MA CAAAJ61359-MA CAAAJ61359-MF AAJ61359-MF
7-Chlorokynurenic acid ≥99%
7-Chlorokynurenic acid
NMDA receptor antagonist at the glycine site

Powder


Formula: C₁₀H₆ClNO₃
MW: 223.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00069227
CAS Number: 18000-24-3
EINECS: 241-913-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty