Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methyl-D-aspartic acid ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Methyl-D-asparaginic AcidMe-D-Asp-OH

J61361-MC J61361-MC J61361-MF J61361-MF J61361-ME J61361-ME
AAJ61361-MFEA 89.47 USD InStock
CAAAJ61361-MC CAAAJ61361-ME CAAAJ61361-MF AAJ61361-MF AAJ61361-ME AAJ61361-MC
N-Methyl-D-aspartic acid ≥98%
N-Methyl-D-aspartic acid
Powder


Formula: C₅H₉NO₄
MW: 147.13 g/mol
Melting Pt: 187…192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004226
CAS Number: 6384-92-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty