Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Guanosine 5'-triphosphate disodium salt

Supplier: Thermo Scientific Chemicals
Synonyms: ((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium saltGuanosine-5-triphosphoric aicd disodium saltGTP disodium salt

J61414-ME J61414-03 J61414-MD J61414-03 J61414-ME J61414-MD
AAJ61414-MDEA 317.36 USD InStock
AAJ61414-MD AAJ61414-ME AAJ61414-03 CAAAJ61414-MD CAAAJ61414-ME CAAAJ61414-03
Guanosine 5'-triphosphate disodium salt
Guanosine 5'-triphosphate disodium salt
Powder


Formula: C₁₀H₁₄N₅Na₂O₁₄P₃
MW: 567.147 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03410297
EINECS: 259-940-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty