Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Histidine monohydrochloride monohydrate cell culture reagent

Supplier: Thermo Scientific Chemicals
Synonyms: H-His-OH.HCl.H2OL-Histidine monohydrochloride monohydrate(S)-(+)-Histidine monohydrochloride monohydrateL-Histidine hydrochloride hydrateHis HCl monohydrate(2S)-(+)-Histidine monohydrochloride monohydrateL-Histidine hydrochloride monohydrateL-3-Imidazol-4-ylalanine monohydrochloride monohydrate

J61465-09 J61465-36 J61465-36 J61465-22 J61465-A1 J61465-09 J61465-22
AAJ61465-22EA 157.52 USD InStock
CAAAJ61465-22 CAAAJ61465-A1 AAJ61465-22 AAJ61465-09 CAAAJ61465-36 CAAAJ61465-09 AAJ61465-36
L(+)-Histidine monohydrochloride monohydrate cell culture reagent
L(+)-Histidine monohydrochloride monohydrate
Powder


Formula: C₆H₉N₃O₂·HCl·H₂O
MW: 209.63 g/mol
Melting Pt: ca 240 °C (dec.)
MDL Number: MFCD00151027
CAS Number: 5934-29-2
Merck Index: 13,04741
https://us-legacy.vwr.com/stibo/web/std.lang.all/98/30/2299830.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty