Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Amygdalin 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: D-Mandelonitrile-β-gentiobiosideLaetrileAmygdalosideAmygdalin(R)-2-Phenyl-2-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)acetonitrileD-Mandelonitrile 6-O-β-D-glucosido-β-D-glucoside

J61472-03 J61472-14 J61472-03 J61472-06 J61472-14 J61472-06
AAJ61472-06EA 90.89 USD InStock
AAJ61472-06 CAAAJ61472-03 CAAAJ61472-14 CAAAJ61472-06 AAJ61472-03 AAJ61472-14
D-Amygdalin 98%
D-Amygdalin
A benzylic glycoside that has been used as an antineoplastic agent


Formula: C₂₀H₂₇NO₁₁
MW: 457.43 g/mol
Melting Pt: 210…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00006598
CAS Number: 29883-15-6
EINECS: 249-925-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty