Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Protease Inhibitor Cocktail, for mammalian Cells

Supplier: Thermo Scientific
J61473-XF J61473-XF
AAJ61473-XFEA 264.78 USD InStock
CAAAJ61473-XF AAJ61473-XF
Protease Inhibitor Cocktail, for mammalian Cells
Cell Culture Supplements and Additives
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00677817

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty