Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bradford's reagent for dye, ready-to-use

Supplier: Thermo Scientific Chemicals
Danger

J61522-K2 J61522-AP J61522-K2 J61522-AP
AAJ61522-APEA 143.1 USD InStock
CAAAJ61522-AP AAJ61522-AP CAAAJ61522-K2 AAJ61522-K2
Bradford's reagent for dye, ready-to-use
Bradford's reagent
Suitable for protein concentration determination (100-1500µg)


Storage Temperature: Refrigerator
UN: 1805
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty