Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aspartame

Supplier: Thermo Scientific
J61523-06 J61523-06 J61523-22 J61523-22 J61523-14 J61523-14
AAJ61523-22EA 420.92 USD InStock
CAAAJ61523-22 AAJ61523-22 AAJ61523-14 CAAAJ61523-06 AAJ61523-06 CAAAJ61523-14
Aspartame
Proteins and Peptides
A non-nutritive sweetener and dipeptide ester about 160 times sweeter than sucrose.

  • EINECS: 245-261-3
  • Melting point: 243 to 245 °C
  • Merck Index: 14,839

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty