Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Metoclopramide hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

J61545-22 J61545-09 J61545-14 J61545-14 J61545-22 J61545-09
AAJ61545-09EA 55.72 USD InStock
CAAAJ61545-22 AAJ61545-09 AAJ61545-14 AAJ61545-22 CAAAJ61545-09 CAAAJ61545-14
Metoclopramide hydrochloride
Metoclopramide hydrochloride
5-HT3 serotonin antagonist; D2 dopamine antagonist

Crystalline solid


Formula: C₁₄H₂₂ClN₃O₂·HCl
MW: 336,26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00058011
CAS Number: 7232-21-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty