Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fluorescein disodium salt

Supplier: Thermo Scientific Chemicals
Synonyms: 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate Fluorescein sodium salt

J61549-22 J61549-36 J61549-22 J61549-36
AAJ61549-22EA 51.3 USD InStock
CAAAJ61549-36 AAJ61549-22 AAJ61549-36 CAAAJ61549-22
Fluorescein disodium salt
Fluorescein disodium salt
A fluorophore comonly used in microscopy

Powder


Formula: C₂₀H₁₀Na₂O₅
MW: 376.28 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00167039
CAS Number: 518-47-8
EINECS: 208-253-0
Merck Index: 13,04185
https://us-legacy.vwr.com/stibo/web/std.lang.all/57/66/2375766.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty