Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dicloxacillin sodium salt monohydrate

Supplier: Thermo Scientific Chemicals
Danger

J61581-03 J61581-03 J61581-06 J61581-06
AAJ61581-06EA 231.28 USD InStock
AAJ61581-06 CAAAJ61581-03 AAJ61581-03 CAAAJ61581-06
Dicloxacillin sodium salt monohydrate
Dicloxacillin sodium salt monohydrate
An antibiotic used in the treatment of staphyloccoci bacteria that are resistant to penicillin G

Powder


Formula: C₁₉H₁₆Cl₂N₃NaO₅S·1H₂O
Melting Pt: 222…225 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00056865
CAS Number: 13412-64-1
EINECS: 603-794-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty