Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) 97%

Supplier: Thermo Scientific Chemicals
Synonyms: ((2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy-((((2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy)hydroxyphosphoryl)oxyphosphinic acid disodium saltNicotinamide adenine dinucleotide reduced, disodium salt (reduced form)

J61638-06 J61638-03 J61638-03 J61638-06
AAJ61638-06EA 454.58 USD InStock
AAJ61638-06 AAJ61638-03 CAAAJ61638-03 CAAAJ61638-06
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) 97%
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)
Powder


Formula: C₂₁H₂₇N₇Na₂O₁₄P₂
MW: 709.41 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036200
CAS Number: 606-68-8
EINECS: 210-123-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty