Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate

Supplier: Thermo Scientific Chemicals
Synonyms: 5′-AMP-Na2 hydrate

J61643-22 J61643-06 J61643-22 J61643-14 J61643-14 J61643-06
AAJ61643-22EA 370.93 USD InStock
CAAAJ61643-14 CAAAJ61643-06 AAJ61643-22 AAJ61643-06 AAJ61643-14 CAAAJ61643-22
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate
Powder


Formula: C₁₀H₁₂N₅Na₂O₇P·H₂O
MW: 391.2 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD00065023
CAS Number: 4578-31-8
EINECS: 224-961-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty