Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lecithin (from soy bean) ≥90%

Supplier: Thermo Scientific Chemicals

J61675-30 J61675-22 J61675-22 J61675-30 J61675-18 J61675-18
AAJ61675-18EA 30.55 USD InStock
AAJ61675-18 CAAAJ61675-30 AAJ61675-22 CAAAJ61675-22 CAAAJ61675-18 AAJ61675-30
Lecithin (from soy bean) ≥90%
Lecithin
Naturally occuring lipid that protects cells from oxidation. Useful as an emulsifier

  • Pale brown to yellow Solid
  • Odourless
  • Beilstein Reference: 5209585


MDL Number: MFCD00147406
CAS Number: 8002-43-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty