Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Histamine (free base)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(1H-Imidazol-4-yl)ethan-1-amine

J61727-03 J61727-03 J61727-06
AAJ61727-03EA 101.22 USD InStock
CAAAJ61727-03 CAAAJ61727-06 AAJ61727-03
Histamine (free base)
Histamine (free base)
Endogenous H1 and H2 histamine receptor agonist; H1 activation mobilizes calcium; H2 activation stimulates adenylate cyclase activity in neurons; activates nitric oxide synthetase; potent vasodilator

Powder


Formula: C₅H₉N₃
MW: 111.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005210
CAS Number: 51-45-6
EINECS: 200-100-6
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty