J61763-03
J61763-06
J61763-06
J61763-03
AAJ61763-03EA
153.58
USD
InStock
CAAAJ61763-03
CAAAJ61763-06
AAJ61763-03
AAJ61763-06
Polymyxin B sulphate, Cell Culture Grade cell culture reagent
Polymyxin B sulphate
Antibiotic for Gram-negative bacteria
White to pale yellow powder
Soluble in water