Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Erioglaucine disodium salt

Supplier: Thermo Scientific Chemicals
Synonyms: Food Blue No.1Brilliant Blue FCFErioglaucin AFD and C Blue No. 1

J61780-06 J61780-14 J61780-14 J61780-06
AAJ61780-14EA 85.84 USD InStock
CAAAJ61780-06 AAJ61780-14 CAAAJ61780-14 AAJ61780-06
Erioglaucine disodium salt
Erioglaucine disodium salt
Powder


Formula: C₃₇H₃₆N₂O₉S₃
MW: 792.84 g/mol
Melting Pt: 283 °C
Storage Temperature: Ambient
MDL Number: MFCD00012141
CAS Number: 3844-45-9
EINECS: 223-339-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty