Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Substance P Free Acid

Supplier: Thermo Scientific
J61828-LB0 J61828-LB0
AAJ61828-LB0EA 116.66 USD InStock
AAJ61828-LB0 CAAAJ61828-LB0
Substance P Free Acid
Proteins and Peptides
CAS No.: 71977-09-8
Molecular Formula: C63H97N17O14S
Formula Weight: 1348.63
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00076781

A neurokinin NK-1 receptor agonist

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty