Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Lactic acid

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (R)-(-)-2-Hydroxypropanoic acid (R)-(-)-2-Hydroxypropionic acid (R)-(-)-Lactic acid

J61871-03 J61871-03 J61871-MD J61871-MD
AAJ61871-MDEA 274.19 USD InStock
CAAAJ61871-03 AAJ61871-MD CAAAJ61871-MD AAJ61871-03
D-(-)-Lactic acid
D(-)-Lactic acid
Useful chiral synthon, building block for depsipeptides

Powder


Formula: C₃H₆O₃
MW: 90.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00068313
CAS Number: 10326-41-7
EINECS: 233-713-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty