Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
Orange G/Blue Loading Dye (6X), Thermo Scientific
Supplier: Thermo Scientific
J61877-AB J61877-AC J61877-AC J61877-AB
AAJ61877-ABEA 110.24 USD InStock
CAAAJ61877-AB AAJ61877-AB CAAAJ61877-AC AAJ61877-AC
Orange G/Blue Loading Dye (6X), Thermo Scientific
Stains Electrophoresis Stains
Orange G/blue loading dye (6X) is used for loading DNA samples into gel electrophoresis wells and tracking migration during electrophoresis.
  • Contains: 0.25% Orange-G, 0.25% bromophenol blue, 0.25% xylene cyanol FF, 15% Ficoll, 30 mM EDTA in autoclaved DI water
It is used in molecular biology protocols and in nucleic acid gel electrophoresis and blotting. It is also used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty