Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium alginate, high viscosity

Supplier: Thermo Scientific Chemicals
Synonyms: Alginic acid sodium salt

J61887-A1 J61887-22 J61887-30 J61887-30 J61887-A1 J61887-22
AAJ61887-A1EA 769.78 USD InStock
AAJ61887-A1 AAJ61887-22 AAJ61887-30 CAAAJ61887-30 CAAAJ61887-A1 CAAAJ61887-22
Sodium alginate, high viscosity
Sodium alginate
A gelling polysaccharide extracted from giant brown seaweed, useful in viscosity studies

Powder


Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty