Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Methionine Cell Culture Reagent

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acidL(-)-MethionineMethionine(S)-(-)-2-Amino-4-(methylthio)butanoic acidH-Met-OH(S)-(-)-Methionine(S)-2-Amino-4-(methylthio)butanoic acid

J61904-36 J61904-18 J61904-18 J61904-36 J61904-22 J61904-22
AAJ61904-22EA 114.17 USD InStock
CAAAJ61904-22 AAJ61904-22 AAJ61904-36 CAAAJ61904-18 AAJ61904-18 CAAAJ61904-36
L-Methionine Cell Culture Reagent
L-Methionine
Powder


Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 284 °C (dec.)
Density: 1.373 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063097
CAS Number: 63-68-3
EINECS: 200-562-9
Merck Index: 13,06004
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty