Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Inosine 5'-monophosphate disodium salt hydrate

Supplier: Thermo Scientific Chemicals
Synonyms: Disodium inosinate hydrate5'-Inosinic acid disodium salt hydrateInosic acid disodium salt hydrate5'-IMP 2Na hydrateInosine 5'-monophosphate disodium salt xhydrate

J61959-22 J61959-14 J61959-22 J61959-14 J61959-06 J61959-06
AAJ61959-14EA 97.17 USD InStock
AAJ61959-14 AAJ61959-06 CAAAJ61959-06 CAAAJ61959-14 CAAAJ61959-22 AAJ61959-22
Inosine 5'-monophosphate disodium salt hydrate
Inosine 5'-monophosphate disodium salt hydrate
Crystalline powder


Formula: C₁₀H₁₁N₄Na₂O₈P·xH₂O
MW: 392.17 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00150372
CAS Number: 352195-40-5
EINECS: 225-146-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty