Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium cholate

Supplier: Thermo Scientific Chemicals
Synonyms: 3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt3α,7α,12α-Trihydroxycholanic acid sodium salt3α,7α,12α-Trihydroxycholan-24-oic acid sodium saltSodium 3α,7α,12α-trihydroxy-5β-cholanoateSodium 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanoateCholic acid sodium salt3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt

J62050-14 J62050-14 J62050-22 J62050-22 J62050-30 J62050-30
AAJ62050-30EA 314.42 USD InStock
CAAAJ62050-30 AAJ62050-30 CAAAJ62050-14 AAJ62050-22 AAJ62050-14 CAAAJ62050-22
Sodium cholate
Sodium cholate
Biochemical solubilizing agent

Powder


Formula: C₂₄H₃₉NaO₅
MW: 430.56 g/mol
Melting Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00150749
CAS Number: 361-09-1
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty