Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dextran sulfate sodium salt, M.W. ∼ 8,000

Supplier: Thermo Scientific Chemicals
Synonyms: Dextran sulfate sodium from DextranDextran sulphate sodium from Dextran

J62101-22 J62101-14 J62101-A1 J62101-22 J62101-14
AAJ62101-14EA 86.93 USD InStock
CAAAJ62101-14 CAAAJ62101-22 AAJ62101-14 AAJ62101-A1 AAJ62101-22
Dextran sulfate sodium salt, M.W. ∼ 8,000
Dextran sulphate sodium salt
Powder


Formula: (C₆H₇Na₃O₁₄S₃)n
Storage Temperature: Ambient
MDL Number: MFCD00081551
CAS Number: 9011-18-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty