Would you like to log in again?
Pig Glutamic pyruvic transaminase (from Heart)
Supplier: Thermo Scientific
J62109-JPL J62109-4I J62109-4I
AAJ62109-JPLEA 269.93 USD InStock
CAAAJ62109-4I AAJ62109-JPL AAJ62109-4I
Pig Glutamic pyruvic transaminase (from Heart)
Enzymes
Applications: Converts L-alanine and alpha-ketoglutarate to pyruvic acid and L-glutamate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty