Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ergocalciferol (vitamin D2, calciferol) 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (S,Z)-3-((E)-2-((1R,3aS,7aR)-1-((2R,5R,E)-5,6-Dimethylhept-3-en-2-yl)-7a-methylhexahydro-1H-inden-4(2H)-ylidene)ethylidene)-4-methylenecyclohexanol

J62163-03 J62163-06 J62163-06 J62163-03
AAJ62163-03EA 51.16 USD InStock
CAAAJ62163-03 CAAAJ62163-06 AAJ62163-03 AAJ62163-06
Ergocalciferol (vitamin D2, calciferol) 98%
Ergocalciferol (Vitamin D2, Calciferol)
Powder


Formula: C₂₈H₄₄O
MW: 396.66 g/mol
Melting Pt: 118…119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00166988
CAS Number: 50-14-6
EINECS: 200-014-9
UN: 2811
ADR: "6.1",III
Merck Index: 14,10018
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty