Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adenosine-3',5'-cyclic monophosphate sodium salt 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 3',5'-Cyclic AMP Sodium Salt3',5'-cAMP Na

J62174-MD J62174-03 J62174-06 J62174-MD J62174-03 J62174-06
AAJ62174-06EA 1028.61 USD InStock
CAAAJ62174-03 CAAAJ62174-06 CAAAJ62174-MD AAJ62174-06 AAJ62174-03 AAJ62174-MD
Adenosine-3',5'-cyclic monophosphate sodium salt 99%
Adenosine-3',5'-cyclic monophosphate sodium salt
Crystalline powder


Formula: C₁₀H₁₁N₅NaO₆P
MW: 351.19 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00069736
CAS Number: 37839-81-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty