Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Serine cell culture reagent

Supplier: Thermo Scientific Chemicals
Synonyms: Serine(S)-(+)-SerineH-Ser-OH(S)-2-Amino-3-hydroxypropanoic acid

J62187-36 J62187-36 J62187-A1 J62187-22 J62187-09 J62187-09 J62187-A1 J62187-22
AAJ62187-A1EA 803.31 USD InStock
CAAAJ62187-36 AAJ62187-A1 AAJ62187-22 AAJ62187-36 CAAAJ62187-09 CAAAJ62187-22 CAAAJ62187-A1 AAJ62187-09
L(+)-Serine cell culture reagent
L(+)-Serine
Powder


Formula: C₃H₇NO₃
MW: 105.09 g/mol
Melting Pt: 222 °C
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS Number: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty