Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Nicotinoylglycine ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: Nicotinuric Acid

J62215-14 J62215-06 J62215-14 J62215-06
AAJ62215-06EA 125.78 USD InStock
AAJ62215-06 AAJ62215-14 CAAAJ62215-14 CAAAJ62215-06
N-Nicotinoylglycine ≥98%
N-Nicotinoylglycine
The major detoxification product of nicotinic acid. A nicotinic acid antagonist

Crystalline powder


Formula: C₈H₈N₂O₃
MW: 180.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023578
CAS Number: 583-08-4
EINECS: 209-497-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty