Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9-cis-Retinoic acid, powder

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 9-cis-RAAlitretinoinNSC-659772Panretin

J62219-LB0 J62219-LB0
AAJ62219-LB0EA 263.77 USD InStock
CAAAJ62219-LB0 AAJ62219-LB0
9-cis-Retinoic acid, powder
9-cis-Retinoic acid
A high affinity ligand for the retinoid X receptor


Formula: C₂₀H₂₈O₂
MW: 300.442 g/mol
Melting Pt: 178…180 °C
Storage Temperature: Freezer
MDL Number: MFCD00270072
CAS Number: 5300-03-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty