Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Lysine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-(+)-2,6-Diaminocaproic acidL(+)-Lysine(S)-2,6-Diaminohexanoic acid(S)-(+)-LysineH-Lys-OH(S)-(+)-2,6-Diaminohexanoic acid

J62225-36 J62225-36 J62225-14 J62225-22 J62225-22 J62225-14
AAJ62225-36EA 651 USD InStock
AAJ62225-36 AAJ62225-14 CAAAJ62225-36 CAAAJ62225-14 CAAAJ62225-22 AAJ62225-22
L-Lysine 98%
L-Lysine
Powder


Formula: C₆H₁₄N₂O₂
MW: 146.19 g/mol
Melting Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00064433
CAS Number: 56-87-1
EINECS: 200-294-2
Merck Index: 13,05656
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty