Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Erythromycin cell culture reagent

Supplier: Thermo Scientific Chemicals
Synonyms: (-)-Erythromycin

J62279-09 J62279-14 J62279-14 J62279-09
AAJ62279-09EA 185.26 USD InStock
AAJ62279-09 CAAAJ62279-09 AAJ62279-14 CAAAJ62279-14
Erythromycin cell culture reagent
Erythromycin
Macrolide antibiotic that inhibits bacterial protein synthesis.

Crystalline powder


Formula: C₃₇H₆₇NO₁₃
MW: 733.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00084654
CAS Number: 114-07-8
EINECS: 204-040-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty