Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium deoxycholate 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium 3α,12α-dihydroxy-5β-cholanoateDeoxycholic acid sodium salt3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt

J62288-30 J62288-30 J62288-14 J62288-14 J62288-22 J62288-22
AAJ62288-14EA 67.35 USD InStock
AAJ62288-14 AAJ62288-22 AAJ62288-30 CAAAJ62288-14 CAAAJ62288-30 CAAAJ62288-22
Sodium deoxycholate 97%
Sodium deoxycholate
Powder


Formula: C₂₄H₃₉NaO₄
MW: 414.56 g/mol
Melting Pt: 357…365 °C
Storage Temperature: Ambient
MDL Number: MFCD00064139
CAS Number: 302-95-4
EINECS: 206-132-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty