Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DNA Marker, Broad Range, Thermo Scientific

Supplier: Thermo Scientific
J62291-XF J62291-S0 J62291-XF J62291-S0
AAJ62291-S0EA 413.94 USD InStock
CAAAJ62291-S0 CAAAJ62291-XF AAJ62291-S0 AAJ62291-XF
DNA Marker, Broad Range, Thermo Scientific
Electrophoresis Reagents Molecular Weight Markers
  • From 250 to 5000 base pairs: 5 bands from 125 to 1000 bp, four bands from 1500 to 5000 bp
  • Recommended load volume is 20 µl
  • Store at room temperature
  • Form: Liquid

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty