Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Cystine dihydrochloride 99%

Supplier: Thermo Scientific Chemicals
Synonyms: (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid) dihydrochloride(2R,2'R)-3,3'-Disulfanediylbis(2-aminopropanoic acid) dihydrochloride

J62292-22 J62292-14 J62292-30 J62292-14 J62292-30 J62292-22
AAJ62292-30EA 492.66 USD InStock
CAAAJ62292-22 CAAAJ62292-14 AAJ62292-30 AAJ62292-14 AAJ62292-22 CAAAJ62292-30
L-Cystine dihydrochloride 99%
L-Cystine dihydrochloride
Powder


Formula: C₆H₁₂N₂O₄S₂·2HCl
MW: 313.23 g/mol
Melting Pt: 228…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00070399
CAS Number: 30925-07-6
EINECS: 250-391-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty