Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Croton oil

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tiglium oil

J62367-09 J62367-09 J62367-22 J62367-14 J62367-14 J62367-22
AAJ62367-14EA 146.07 USD InStock
CAAAJ62367-14 AAJ62367-14 CAAAJ62367-22 AAJ62367-09 CAAAJ62367-09 AAJ62367-22
Croton oil
Croton oil
A natural source of phorbol, phorbol esters and glycerides of saturated fatty acids

Oil


Flash Pt: 74 °C(165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00130880
CAS Number: 8001-28-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty