Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Blocking Reagents, Denhardt's Solution (100X), Thermo Scientific

Supplier: Thermo Scientific
J62416-AE J62416-AE J62416-AD J62416-AD
AAJ62416-ADEA 275.26 USD InStock
AAJ62416-AD AAJ62416-AE CAAAJ62416-AE CAAAJ62416-AD
Blocking Reagents, Denhardt's Solution (100X), Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers
Denhardt's solution is used as a blocking reagent for northern or southern blots to prevent probe binding on open membrane space and non-target nucleic acids.

  • Contains 2% ficoll (type 400, pharmacia), 2% polyvinylpyrrolidone and 2% bovine serum albumin
  • Soluble in water

Denhardt's reagent is recommended for northern hybridization, hybridization of RNA, single copy southern hybridization or hybridization of DNA, immobilized on nylon membranes.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty